Lithium perchlorate trihydrate 99%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

194720250 194722500 194720010
ACRO194720250EA 28.3 EUR
ACRO194720250 ACRO194722500 ACRO194720010
Lithium perchlorate trihydrate 99%, extra pure
Lithium perchlorate trihydrate
Formule: ClLiO₄·3H₂O
MW: 160.44 g/mol
Smeltpunt: 95 °C
Dichtheid: 1.841
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149765
CAS nummer: 13453-78-6
EINECS: 232-237-2
UN: 1481
ADR: 5.1 II

Order Now

Specification Test Results

Appearance White Adhering crystals and/or chunks
Titration Argentometric ≥98.5 %
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR