Copper(I) iodide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

194900010 194902500 194900050
ACRO194900010EA 260 EUR
ACRO194900010 ACRO194902500 ACRO194900050
Copper(I) iodide 98%
Copper(I) iodide
Formule: CuI
Kookpunt: 1290 °C (1013 hPa)
Smeltpunt: 605 °C
Dichtheid: 5,62 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010978
CAS nummer: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9,III
Merck Index: 13,02692

Order Now

Specification Test Results

Appearance Off-white to grey-brown Powder
Titration Complexometric ≥97.5 %
Sulfate (SO4) ≤5000 ppm
Iron (Fe) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR