Diammonium tetrachloropalladate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

195170020 195170100
ACRO195170020EA 244 EUR
ACRO195170020 ACRO195170100
Diammonium tetrachloropalladate 98%
Diammonium tetrachloropalladate
Formule: H₈Cl₄N₂Pd
Dichtheid: 2,17 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010884
CAS nummer: 13820-40-1
EINECS: 237-498-6

Order Now

Specification Test Results

Appearance Green to red to brown Powder or crystals
Palladium (Pd) ≥36.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR