Copper(II) carbonate basic Cu 54-56%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

196670250 196675000 196670025
ACRO196670250EA 21.6 EUR
ACRO196670250 ACRO196675000 ACRO196670025
Copper(II) carbonate basic Cu 54-56%, pure
Copper(II) carbonate basic
Formule: CuCO₃·Cu(OH)₂
Smeltpunt: 200 °C
Dichtheid: 4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010976
CAS nummer: 12069-69-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Green Crystalline powder
Assay 54 to 56 % (Cu)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR