Koper(II)sulfaat pentahydraat 98%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

197730010 197730050 197730250
ACRO197730010EA 45.4 EUR
ACRO197730010 ACRO197730050 ACRO197730250
Koper(II)sulfaat pentahydraat 98%, extra pure
Koper(II)sulfaat pentahydraat
Formule: CuSO₄·5H₂O
Dichtheid: 2,284 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149681
CAS nummer: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
Bereik: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Appearance Blue Crystalline powder or crystals
Titration Iodometric ≥97.5 %
Arsenic (As) ≤10 ppm
Iron (Fe) ≤200 ppm
Lead (Pb) ≤125 ppm
Nickel (Ni) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR