Boortribromide 1 M in methylene chloride

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

198900100 198901000 198908000
ACRO198900100EA 59.6 EUR
ACRO198900100 ACRO198901000 ACRO198908000
Boortribromide 1 M in methylene chloride
Boortribromide
Formule: BBr₃
MW: 250.52 g/mol
Kookpunt: 91.2
Smeltpunt: -46
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011312
CAS nummer: 10294-33-4
EINECS: 233-657-9
UN: 3390
ADR: 6.1
Merck Index: 13,01337

Order Now

Specification Test Results

Appearance Clear yellow to dark brown Solution (May darken on storage. May develop some turbidity or precipitate)
Strength 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR