4-Fluoro-3-nitrobenzotrifluoride 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

200610050 200611000
ACRO200610050EA 18.4 EUR
ACRO200610050 ACRO200611000
4-Fluoro-3-nitrobenzotrifluoride 96%
4-Fluoro-3-nitrobenzotrifluoride
Formule: C₇H₃F₄NO₂
Kookpunt: 92 °C (15 mmHg)
Dichtheid: 1,49 g/cm³
Vlampunt: 33 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007059
CAS nummer: 367-86-2
EINECS: 206-702-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR