Ammonium iron(II) sulphate hexahydrate 99+% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate

201370010 201370250 201375000 201370050 201370251
ACRO201370010EA 72.2 EUR
ACRO201370010 ACRO201370250 ACRO201375000 ACRO201370050 ACRO201370251
Ammonium iron(II) sulphate hexahydrate 99+% for analysis
Ammonium iron(II) sulphate hexahydrate
Formule: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Smeltpunt: 100 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150530
CAS nummer: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Appearance Light green Crystalline powder or crystals
Titration with KMnO4 ≥99.0 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤30 ppm
Copper (Cu) ≤20 ppm
Manganese (Mn) ≤500 ppm
Zinc (Zn) ≤30 ppm
Ferric iron (Fe 3+) ≤200 ppm (May increase due to inherent oxidation on storage)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR