Diammonium hydrogen orthophosphate 99+% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

201825000 201822500 201820025
ACRO201825000EA 64.3 EUR
ACRO201825000 ACRO201822500 ACRO201820025
Diammonium hydrogen orthophosphate 99+% for analysis
Diammonium hydrogen orthophosphate
Formule: (NH₄)₂HPO₄
Smeltpunt: 155 °C
Dichtheid: 1,619 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010891
CAS nummer: 7783-28-0
EINECS: 231-987-8
Bereik: 01-2119490974-22
Merck Index: 13,00546

Order Now

Specification Test Results

Appearance White Adhering crystals
pH 7.9 to 8.3 (5 % at 20°C)
Titration with HCl ≥99.0 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤10 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR