Potassium selenocyanate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

201980100 201980500 201982500
ACRO201980100EA 90.2 EUR
ACRO201980100 ACRO201980500 ACRO201982500
Potassium selenocyanate 99%
Potassium selenocyanate
Formule: CKNSe
Smeltpunt: 152…158 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011366
CAS nummer: 3425-46-5
EINECS: 222-320-1
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR