Zinc bromide, watervrij 98%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

202101000 202105000 202100025
ACRO202101000EA 43.5 EUR
ACRO202101000 ACRO202105000 ACRO202100025
Zinc bromide, watervrij 98%, extra pure
Zinc bromide
Formule: Br₂Zn
Kookpunt: 697 °C (760 mmHg)
Smeltpunt: 394 °C
Dichtheid: 4,22 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011294
CAS nummer: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 13,10182

Order Now

Specification Test Results

Appearance White to off-white Powder and/or chunks
Titration Argentometric ≥97.5 %
Water ≤2 % (K.F.)
Sulfate (SO4) ≤100 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤50 ppm
Magnesium (Mg) ≤100 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR