Amberlite® CG-50 resin_ion-exchange Type 1

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

202110250 202115000 202111000
ACRO202110250EA 62.2 EUR
ACRO202110250 ACRO202115000 ACRO202111000
Amberlite® CG-50 resin_ion-exchange Type 1
Amberlite® CG-50

CAS No.: 9042-11-9

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00132693
CAS nummer: 9042-11-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size ≤10 % (> 150 µm)
Moisture ≤10 %
Total weight capacity ≥10 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR