6-Aminonicotinic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

202540050 202540250 202540010
ACRO202540050EA 91 EUR
ACRO202540050 ACRO202540250 ACRO202540010
6-Aminonicotinic acid 98%
6-Aminonicotinic acid
Formule: C₆H₆N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006326
CAS nummer: 3167-49-5
EINECS: 221-630-4
Merck Index: 13,00455

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR