Potassium hexacyanocobaltate(III) 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Tripotassium hexacyanocobaltate(III)

203920050 203921000 203925000
ACRO203920050EA 19.7 EUR
ACRO203920050 ACRO203921000 ACRO203925000
Potassium hexacyanocobaltate(III) 95%
Potassium hexacyanocobaltate(III)

Appearance: Light yellow to light brown Crystalline solid

Formule: C₆CoK₃N₆
MW: 332.332 g/mol
Dichtheid: 1.87 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011391
CAS nummer: 13963-58-1

Order Now

Specification Test Results

Appearance Light yellow to light brown Crystalline solid
Assay ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR