Ammonium heptamolybdate tetrahydrate 99+% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

205851000 205855000 205850025
ACRO205851000EA 43.9 EUR
ACRO205851000 ACRO205855000 ACRO205850025
Ammonium heptamolybdate tetrahydrate 99+% for analysis
Ammonium heptamolybdate tetrahydrate
Formule: (NH₄)₆Mo₇O₂₄·4H₂O
Smeltpunt: 190 °C
Dichtheid: 2.49 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167059
CAS nummer: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay ≥99.0 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤100 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR