Ammonium nitrate 99+% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

205860010 205865000 205861000 205860025
ACRO205860010EA 91.9 EUR
ACRO205860010 ACRO205865000 ACRO205861000 ACRO205860025
Ammonium nitrate 99+% for analysis
Ammonium nitrate
Formule: NH₄NO₃
Kookpunt: 210 °C (760 mmHg)
Smeltpunt: 169 °C
Dichtheid: 1,72 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011425
CAS nummer: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1,III
Merck Index: 13,00538

Order Now

Specification Test Results

Appearance White Adhering crystals
Residue after ignition ≤0.01 % (as SO4)
pH 4.5 to 6 (5 % at 20°C)
Titration with NaOH ≥99.0 %
Chloride (Cl) ≤5 ppm
Nitrite (NO2) ≤5 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR