Ammonium sulphate 99.5% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

205870010 205870050 205870250 205872500
ACRO205870010EA 48 EUR
ACRO205870010 ACRO205870050 ACRO205870250 ACRO205872500
Ammonium sulphate 99.5% for analysis
Ammonium sulphate
Formule: (NH₄)₂SO₄
Smeltpunt: 280 °C
Dichtheid: 1,78 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003391
CAS nummer: 7783-20-2
EINECS: 231-984-1
Bereik: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Appearance Colorless to white Crystals
Residue after ignition ≤0.01 % (as SO4)
pH 5 to 6 (5 % at 20°C)
Titration excess NaOH ≥99.5 %
Chloride (Cl) ≤3 ppm
Nitrate (NO3) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤10 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR