1-Octanesulphonic acid sodium salt monohydrate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate

206140050 206140250 206141000
ACRO206140050EA 70.6 EUR
ACRO206140050 ACRO206140250 ACRO206141000
1-Octanesulphonic acid sodium salt monohydrate 98%
1-Octanesulphonic acid sodium salt monohydrate
Formule: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007544
CAS nummer: 207596-29-0
EINECS: 226-195-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying 7 % to 9 % (140°C)
Titration after Ion exchange ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR