Lithium iodide hydrate 99%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

208011000 208015000
ACRO208011000EA 73.2 EUR
ACRO208011000 ACRO208015000
Lithium iodide hydrate 99%, extra pure
Lithium iodide hydrate
Formule: ILi·xH₂O
Kookpunt: 1171 °C (760 mmHg)
Smeltpunt: 73 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149769
CAS nummer: 85017-80-7
EINECS: 233-822-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤30 % (K.F.)
Sulfate (SO4) ≤1000 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤15 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR