N-Methyl-bis-trifluoroacetamide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

210070050 210070250
ACRO210070050EA 196 EUR
ACRO210070050 ACRO210070250
N-Methyl-bis-trifluoroacetamide 98%
N-Methylbistrifluoroacetamide
Formule: C₅H₃F₆NO₂
Kookpunt: 120…122 °C (760 mmHg)
Dichtheid: 1,547 g/cm³
Vlampunt: 45 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000412
CAS nummer: 685-27-8
EINECS: 211-680-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3450 to 1.3470 (20°C, 589 nm)
GC ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR