O-Methylhydroxylamine hydrochloride 98+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

210490050 210490250 210491000
ACRO210490050EA 90.3 EUR
ACRO210490050 ACRO210490250 ACRO210491000
O-Methylhydroxylamine hydrochloride 98+%
O-Methylhydroxylamine hydrochloride
Formule: CH₆ClNO
Smeltpunt: 148…153 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012951
CAS nummer: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 153 °C
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR