Potassium hexacyanoferrate(II) trihydrate 99+% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate

211095000 211090025
ACRO211095000EA 56.5 EUR
ACRO211095000 ACRO211090025
Potassium hexacyanoferrate(II) trihydrate 99+% for analysis
Potassium hexacyanoferrate(II) trihydrate
Formule: K₄Fe(CN)₆·3H₂O
Smeltpunt: 70 °C
Dichtheid: 1,85 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167023
CAS nummer: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Titration with KMnO4 ≥99.0 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR