N-(Benzyloxycarbonyloxy)succinimide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

211170250 211171000 211175000
ACRO211170250EA 51.3 EUR
ACRO211170250 ACRO211171000 ACRO211175000
N-(Benzyloxycarbonyloxy)succinimide 98%
N-(Benzyloxycarbonyloxy)succinimide
Formule: C₁₂H₁₁NO₅
Smeltpunt: 77…82 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005513
CAS nummer: 13139-17-8
EINECS: 236-075-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 82 °C
Loss on drying ≤0.25 % (60 °C, 2 h, 1 mmHg)
Solubility (10 % in acetone) Clear colorless to almost colorless
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR