Purpurin(1,2,4-trihydroxy-9,10-anthraquinone)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

211990250 211990050
ACRO211990250EA 158 EUR
ACRO211990250 ACRO211990050
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone)
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone)
Formule: C₁₄H₈O₅
Smeltpunt: 253…256 °C
Vlampunt: 113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001203
CAS nummer: 81-54-9
EINECS: 201-359-8

Order Now

Specification Test Results

Appearance Dark red to orange-brown to brown Powder
Infrared spectrum Conforms
Solubility (1 mg/ml in NH4OH) Red to very dark red to purple
HPLC ≥85 %
UV Lambda max 470 to 490 nm (10 mg/l methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR