Tetrabutylammonium tetrahydroborate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate

212900100 212900500
ACRO212900100EA 72.2 EUR
ACRO212900100 ACRO212900500
Tetrabutylammonium tetrahydroborate 98%
Tetrabutylammonium tetrahydroborate
Formule: C₁₆H₄₀BN
MW: 257.31 g/mol
Smeltpunt: 126…131 °C
MDL Nummer: MFCD00012035
CAS nummer: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3 II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR