Methyl-(S)-(-)-lactaat 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (S)-Methyl 2-hydroxypropanoate, Methyl (S)-(-)-2-hydroxypropionate, Sodium carboxymethyl cellulose

213060010 213062500 213060025
ACRO213060010EA 235 EUR
ACRO213060010 ACRO213062500 ACRO213060025
Methyl-(S)-(-)-lactaat 97%
Methyl-(S)-(-)-lactaat
Formule: C₄H₈O₃
MW: 104.11 g/mol
Kookpunt: 144
Smeltpunt: –44 °C
Dichtheid: 1.092 g/cm³
Vlampunt: 55 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064265
CAS nummer: 27871-49-4
EINECS: 248-704-9
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4160 (20°C, 589 nm)
Density (20°C) 1.085 to 1.098 g/ml
GC ≥96.0 %
Specific optical rotation -8° to -9° (20°C, 589 nm) (neat)
Free acid ≤1.0 % (lactic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR