Dikwikdichloride 99+%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

213120050 213121000 213125000
ACRO213120050EA 36.1 EUR
ACRO213120050 ACRO213121000 ACRO213125000
Dikwikdichloride 99+%, extra pure
Dikwikdichloride
Formule: Cl₂Hg₂
Kookpunt: 384 °C (1013 hPa)
Dichtheid: 7,15 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011043
CAS nummer: 10112-91-1
EINECS: 233-307-5
UN: 2025
ADR: 6.1,III
Merck Index: 12,05951

Order Now

Specification Test Results

Appearance Off-white Powder
Titration Iodimetric ≥99.0 %
Mercuric Chloride ≤0.01 % (HgCl2)
Residue after reduction ≤0.02 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR