Mangaandioxide 80-85%, powder

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

213490010 213490250 213490025
ACRO213490010EA 0 EUR
ACRO213490010 ACRO213490250 ACRO213490025
Mangaandioxide 80-85%, powder
Mangaandioxide
Formule: MnO₂
MW: 86.94 g/mol
Smeltpunt: 535 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003463
CAS nummer: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730

Order Now

Specification Test Results

Appearance Dark grey Powder
Loss on drying ≤0.5 % (105°C)
Titration with Na2S2O3 ≥80 %
Silicate (as SiO2) ≤7 %
Iron (Fe) ≤1 %
Insoluble matter ≤10 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR