HEPES sodium salt 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, HEPES sodium, HEPES Na

215000250 215001000 215000010 215005000
ACRO215000250EA 50.9 EUR
ACRO215000250 ACRO215001000 ACRO215000010 ACRO215005000
HEPES sodium salt 99%
HEPES sodium salt
Formule: C₈H₁₇N₂NaO₄S
Smeltpunt: 248 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036463
CAS nummer: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤3 % (K.F.)
UV at 260 nm A: ≤0.04
pH 9.5 to 10.5 (1 % in water)
Titration with HClO4 ≥98.5 % (on anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR