Methylenediphosphonic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Medronic acid

215070010 215070050
ACRO215070010EA 62 EUR
ACRO215070010 ACRO215070050
Methylenediphosphonic acid 98%
Methylenediphosphonic acid
Formule: CH₆O₆P₂
MW: 176 g/mol
Kookpunt: 82 °C (6 mmHg)
Smeltpunt: 200…204 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002139
CAS nummer: 1984-15-2
EINECS: 217-851-0
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 204 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR