Benzyl cinnamate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

215291000 215295000 215290050
ACRO215291000EA 39.9 EUR
ACRO215291000 ACRO215295000 ACRO215290050
Benzyl cinnamate 99%
Benzyl cinnamate
Formule: C₁₆H₁₄O₂
Kookpunt: 195…200 °C (5 mmHg)
Smeltpunt: 33…37 °C
Vlampunt: >112 °C
MDL Nummer: MFCD00004789
CAS nummer: 103-41-3
EINECS: 203-109-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Crystalline mass or liquid after melting
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥98.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR