Methyl laurate 98+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

215560025 215560100 215560500 215562500
ACRO215560025EA 24.6 EUR
ACRO215560025 ACRO215560100 ACRO215560500 ACRO215562500
Methyl laurate 98+%
Methyl laurate
Formule: C₁₃H₂₆O₂
Kookpunt: 262 °C (766 mmHg)
Smeltpunt: 4…5 °C
Dichtheid: 0,87 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008966
CAS nummer: 111-82-0
EINECS: 203-911-3
UN: 3082

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4340 (20°C, 589 nm)
GC ≥98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR