N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

216531000 216535000
ACRO216531000EA 48.5 EUR
ACRO216531000 ACRO216535000
N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%
N-(3-(Trimethoxysilyl)propyl)ethylenediamine
Formule: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
Kookpunt: 261…263 °C (1013 hPa)
Smeltpunt: ∼ 0 °C
Dichtheid: 1,03 g/cm³
Vlampunt: 136 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008173
CAS nummer: 1760-24-3
EINECS: 217-164-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4425 to 1.4445 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR