Aluminiumkaliumsulfaat dodecahydraat 99.5% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

217480010 217485000 217480025
ACRO217480010EA 54.5 EUR
ACRO217480010 ACRO217485000 ACRO217480025
Aluminiumkaliumsulfaat dodecahydraat 99.5% for analysis
Aluminium potassium sulphate dodecahydrate
Formule: AlK(SO₄)₂·12H₂O
Smeltpunt: 92 °C
Dichtheid: 1,72 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149143
CAS nummer: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric ≥99.5 %
pH 3 to 3.5 (10 % at 20°C)
Chloride (Cl) ≤50 ppm
Ammonium (NH4) ≤2000 ppm
Arsenic (As) ≤2 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR