Lead(IV) oxide 97+%, extra pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

217530050 217531000 217535000 217530025
ACRO217530050EA 46.3 EUR
ACRO217530050 ACRO217531000 ACRO217535000 ACRO217530025
Lead(IV) oxide 97+%, extra pure
Lead(IV) oxide
Formule: O₂Pb
Dichtheid: 9,4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011165
CAS nummer: 1309-60-0
EINECS: 215-174-5
UN: 1872
ADR: 5.1,III
Merck Index: 12,05424

Order Now

Specification Test Results

Appearance Brown to black Powder
Titration with Na2S2O3 ≥97.0 %
Chloride (Cl) ≤0.02 %
Sulfate (SO4) ≤0.005 %
Copper (Cu) ≤0.01 %
Iron (Fe) ≤0.01 %
Zinc (Zn) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR