Potassium hydrogen carbonate 99.7+% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassium bicarbonate

217590010 217590051
ACRO217590010EA 96.3 EUR
ACRO217590010 ACRO217590051
Potassium hydrogen carbonate 99.7+% for analysis
Potassium hydrogen carbonate
Formule: KHCO₃
MW: 100.12 g/mol
Smeltpunt: 292 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011402
CAS nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with HCl 99.7 to 100.3 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤10 ppm
Iron (Fe) ≤5 ppm
Sodium (Na) ≤300 ppm
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.005 % (in water)
Nitrogen compounds (as N) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR