Ammonium trifluoroacetate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

218860250 218861000
ACRO218860250EA 49.3 EUR
ACRO218860250 ACRO218861000
Ammonium trifluoroacetate 98%
Ammonium trifluoroacetate
Formule: C₂H₄F₃NO₂
Smeltpunt: 122…126 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012615
CAS nummer: 3336-58-1
EINECS: 222-072-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 122 to 126 °C
Titration with HClO4 ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR