(S)-(+)-1-Aminopropan-2-ol 98+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

218980010 218980050 218980250
ACRO218980010EA 58.6 EUR
ACRO218980010 ACRO218980050 ACRO218980250
(S)-(+)-1-Aminopropan-2-ol 98+%
(S)-(+)-1-Aminopropan-2-ol
Formule: C₃H₉NO
Kookpunt: 160 °C (760 mmHg)
Smeltpunt: 19…21 °C
Dichtheid: 0,954 g/cm³
Vlampunt: 71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064429
CAS nummer: 2799-17-9
EINECS: 220-533-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4465 to 1.4495 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation +16° to +20° (20°C, 589 nm) (c=1.8, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR