Sodium sulphite, anhydrous 98.5% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

219270010 219270250 219270050 219272500
ACRO219270010EA 59.6 EUR
ACRO219270010 ACRO219270250 ACRO219270050 ACRO219272500
Sodium sulphite, anhydrous 98.5% for analysis
Sodium sulphite
Formule: Na₂SO₃
MW: 126.04 g/mol
Smeltpunt: >500 °C (ontbinding)
Dichtheid: 2.63 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003503
CAS nummer: 7757-83-7
EINECS: 231-821-4
Bereik: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Iodimetric ≥98.0 %
Chloride (Cl) ≤100 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR