Kaliumperchloraat 99+% analysis_for

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

221360250 221365000 221360010 221360025
ACRO221360250EA 25.4 EUR
ACRO221360250 ACRO221365000 ACRO221360010 ACRO221360025
Kaliumperchloraat 99+% analysis_for
Kaliumperchloraat
Formule: ClKO₄
MW: 138.55 g/mol
Smeltpunt: 400 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011362
CAS nummer: 7778-74-7
EINECS: 231-912-9
UN: 1489
ADR: 5.1 II
Merck Index: 13,07738

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration ≥99.0 %
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤10 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Sodium (Na) ≤200 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR