Mangaandioxide 99% -10 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

222581000 222585000 222580025
ACRO222581000EA 35.2 EUR
ACRO222581000 ACRO222585000 ACRO222580025
Mangaandioxide 99% -10 mesh
Mangaandioxide
Formule: MnO₂
MW: 86.94 g/mol
Smeltpunt: 535 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003463
CAS nummer: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730

Order Now

Specification Test Results

Appearance Grey to black Powder
Titration with Na2S2O3 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR