Zink, granular 20 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

222601000 222605000 222600025
ACRO222601000EA 24.5 EUR
ACRO222601000 ACRO222605000 ACRO222600025
Zink, granular 20 mesh
Zink
Formule: Zn
Kookpunt: 908 °C (1013 hPa)
Smeltpunt: 420 °C
Dichtheid: 7,14 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
UN: 1436
Merck Index: 12,10255

Order Now

Specification Test Results

Appearance Silver grey Granules or filings
Titration Complexometric ≥99.8 %
Particle size 20 mesh
Suitability (For determination of As) Passes test
Iron (Fe) ≤0.01 %
Lead (Pb) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR