Tin(II) chloride dihydrate 98+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Stannous chloride dihydrate

222661000 222665000 222660025
ACRO222661000EA 32.6 EUR
ACRO222661000 ACRO222665000 ACRO222660025
Tin(II) chloride dihydrate 98+%
Tin(II) chloride dihydrate
Formule: SnCl₂·2H₂O
Kookpunt: 652 °C (760 mmHg)
Smeltpunt: 37…38 °C
Dichtheid: 2,71 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149863
CAS nummer: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals and/or chunks
Titration Iodimetric 98.0 to 103.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR