Sodium perborate tetrahydrate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Sodium peroxometaborate tetrahydrate

223200250 223200010 223200050 223202500
ACRO223200250EA 22.1 EUR
ACRO223200250 ACRO223200010 ACRO223200050 ACRO223202500
Sodium perborate tetrahydrate 97%
Sodium perborate tetrahydrate
Formule: NaH₂BO₄·4H₂O
Smeltpunt: 60 °C
Dichtheid: 1,73 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149231
CAS nummer: 10486-00-7
EINECS: 239-172-9

Order Now

Specification Test Results

Appearance White Crystalline powder
pH 10.1 to 10.6 (1 % solution)
Titration with KMnO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR