Antimony (III) sulphide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

223595000 223591000
ACRO223595000EA 110 EUR
ACRO223595000 ACRO223591000
Antimony (III) sulphide 98%
Antimony (III) sulphide
Formule: S₃Sb₂
Kookpunt: 1150 °C (1013 hPa)
Smeltpunt: 550 °C
Dichtheid: 4,12 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011217
CAS nummer: 1345-04-6
EINECS: 215-713-4
ADR: 6.1,III
Merck Index: 12,00754

Order Now

Specification Test Results

Appearance Dark grey to black Crystalline powder
Assay ≥97.5 % (Sb2S3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR