1-Iodo-2-methylpropane 97% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

223740250 223741000 223745000
ACRO223740250EA 64 EUR
ACRO223740250 ACRO223741000 ACRO223745000
1-Iodo-2-methylpropane 97% stabilised
1-Iodo-2-methylpropane
Formule: C₄H₉I
Kookpunt: 120…121 °C (760 mmHg)
Smeltpunt: –93 °C
Dichtheid: 1,59 g/cm³
Vlampunt: 12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001084
CAS nummer: 513-38-2
EINECS: 208-160-5
UN: 2391
ADR: 3,II
Merck Index: 13,05160

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR