Kalium 98%, chunks in mineral oil

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

223860100 223861000 223860500 223865000
ACRO223860100EA 44.9 EUR
ACRO223860100 ACRO223861000 ACRO223860500 ACRO223865000
Kalium 98%, chunks in mineral oil
Kalium
Formule: K
Kookpunt: 770 °C (760 mmHg)
Smeltpunt: 63 °C
Dichtheid: 0,86 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133776
CAS nummer: 7440-09-7
EINECS: 231-119-8
UN: 2257
ADR: 4.3,I
Merck Index: 12,07763

Order Now

Specification Test Results

Appearance Grey Chunks
Total trace metal impurities ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR