4-tert-Butyl-1-bromobenzene 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

225190250 225191000 225190050
ACRO225190250EA 64.1 EUR
ACRO225190250 ACRO225191000 ACRO225190050
4-tert-Butyl-1-bromobenzene 97%
4-tert-Butyl-1-bromobenzene
Formule: C₁₀H₁₃Br
Kookpunt: 80…81 °C (2 mmHg)
Smeltpunt: 15…16 °C
Dichtheid: 1,22 g/cm³
Vlampunt: 97 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000108
CAS nummer: 3972-65-4
EINECS: 223-599-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5315 to 1.5345 (20°C, 589 nm)
GC ≥96.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR