L-Glutamic acid 5-benzyl ester 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

225670050 225670010 225670250
ACRO225670050EA 65 EUR
ACRO225670050 ACRO225670010 ACRO225670250
L-Glutamic acid 5-benzyl ester 99%
L-Glutamic acid 5-benzyl ester
Formule: C₁₂H₁₅NO₄
Smeltpunt: 162…167 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002633
CAS nummer: 1676-73-9
EINECS: 216-826-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +26° to +29° (20°C, 589 nm) (c=2, 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR