L-Norvaline 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

226020010 226020050 226020500
ACRO226020010EA 32.5 EUR
ACRO226020010 ACRO226020050 ACRO226020500
L-Norvaline 99%
L-Norvaline
Formule: C₅H₁₁NO₂
Kookpunt: 228 °C (1013 hPa)
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064421
CAS nummer: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Specific optical rotation +22.0° to +26.0° (20°C, 589 nm) (c=2, 5 N HCl)
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR